2-Chloro-4-ethoxyphenylboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: 2-Chloro-4-ethoxybenzeneboronic acid

C3222-200MG C3222-1G
TCIAC3222-200MGEA 35 EUR
TCIAC3222-200MG TCIAC3222-1G
2-Chloro-4-ethoxyphenylboronic acid (contains varying amounts of Anhydride)
2-Chloro-4-ethoxyphenylboronic acid
Formula: C₈H₁₀BClO₃
MW: 200,43 g/mol
Número MDL: MFCD02684318
Núm. CAS: 313545-44-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed1eb613b7921966c6a263aab5e2da78e315e74f&vid=ed1eb613b7921966c6a263aab5e2da78e315e74f&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR