2-Chloro-(4-methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-Chloro-4-(methoxycarbonyl)phenylboronic acid, 2-Chloro-4-(methoxycarbonyl)benzeneboronic acid, (2-Chloro-4-(methoxycarbonyl)phenyl)boronic acid
C3224-1G
C3224-5G
TCIAC3224-1GEA
59
EUR
TCIAC3224-1G
TCIAC3224-5G
2-Chloro-(4-methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
2-Chloro-(4-methoxycarbonyl)phenylboronic acid
Formula:
C₈H₈BClO₄ MW: 214,4107 g/mol |
Número MDL:
MFCD08689530 Núm. CAS: 603122-80-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...