5-Carboxy-2-chlorophenylboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-Chloro-5-carboxyphenylboronic acid
C3246-1G
C3246-5G
TCIAC3246-1GEA
53
EUR
TCIAC3246-1G
TCIAC3246-5G
5-Carboxy-2-chlorophenylboronic acid (contains varying amounts of Anhydride)
5-Carboxy-2-chlorophenylboronic acid
Formula:
C₇H₆BClO₄ MW: 200,3800 g/mol |
Número MDL:
MFCD08689491 Núm. CAS: 913835-75-3 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...