4-Chloro-2-ethoxyphenylboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: 4-Chloro-2-ethoxybenzeneboronic acid

C3267-200MG C3267-1G
TCIAC3267-200MGEA 18 EUR
TCIAC3267-200MG TCIAC3267-1G
4-Chloro-2-ethoxyphenylboronic acid (contains varying amounts of Anhydride)
4-Chloro-2-ethoxyphenylboronic acid
Formula: C₈H₁₀BClO₃
MW: 200,43 g/mol
Número MDL: MFCD03094940
Núm. CAS: 850568-80-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0b0857eb0d0fb68e3cd3bf9a4207b19e444af12&vid=b0b0857eb0d0fb68e3cd3bf9a4207b19e444af12&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR