3-Chloro-(4-methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: (3-Chloro-4-(methoxycarbonyl)phenyl)boronic acid, 3-Chloro-4-(methoxycarbonyl)phenylboronic acid, 3-Chloro-4-(methoxycarbonyl)benzeneboronic acid

C3279-1G C3279-5G
TCIAC3279-1GEA 77 EUR
TCIAC3279-1G TCIAC3279-5G
3-Chloro-(4-methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
3-Chloro-(4-methoxycarbonyl)phenylboronic acid
Formula: C₈H₈BClO₄
MW: 214,4100 g/mol
Número MDL: MFCD06659874
Núm. CAS: 603122-82-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4f31663711f016efa68543506d0a59c464b5de0&vid=e4f31663711f016efa68543506d0a59c464b5de0&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR