4-Chloro-3-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-Chloro-m-anisic acid
C3304-1G
C3304-5G
TCIAC3304-1GEA
47
EUR
TCIAC3304-1G
TCIAC3304-5G
4-Chloro-3-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
4-Chloro-3-methoxybenzoic acid
Formula:
C₈H₇ClO₃ MW: 186,5900 g/mol |
Número MDL:
MFCD00269640 Núm. CAS: 85740-98-3 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 217.0 to 221.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...