2-(4-Chlorophenoxy)benzaldehyde ≥98.0% (por GC)
Proveedor: TCI
C3351-1G
C3351-5G
TCIAC3351-1GEA
56
EUR
TCIAC3351-1G
TCIAC3351-5G
2-(4-Chlorophenoxy)benzaldehyde ≥98.0% (por GC)
2-(4-Chlorophenoxy)benzaldehyde
Formula:
C₁₃H₉ClO₂ MW: 232,6600 g/mol |
Número MDL:
MFCD01568734 Núm. CAS: 111826-11-0 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 55.0 to 59.0 deg-C |
Learn more
![](/m-es.vwr.com/es_ES/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...