3-Chloropyrazine-2-carboxamide ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI

C3376-200MG C3376-1G
TCIAC3376-200MGEA 37 EUR
TCIAC3376-200MG TCIAC3376-1G
3-Chloropyrazine-2-carboxamide ≥98.0% (by HPLC, total nitrogen)
3-Chloropyrazine-2-carboxamide
Formula: C₅H₄ClN₃O
MW: 157,56 g/mol
Número MDL: MFCD09863915
Núm. CAS: 21279-62-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a03c59ffa6dcf26de80f2cca202364dd7795a719&vid=a03c59ffa6dcf26de80f2cca202364dd7795a719&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 187.0 to 191.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR