2-Chloro-5-fluoropyrimidin-4-one ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
2-Chloro-5-fluoro-4-pyrimidinone
C3381-1G
C3381-5G
TCIAC3381-1GEA
67
EUR
TCIAC3381-1G
TCIAC3381-5G
2-Chloro-5-fluoropyrimidin-4-one ≥98.0% (by titrimetric analysis)
2-Chloro-5-fluoropyrimidin-4-one
Formula:
C₄H₂ClFN₂O MW: 148,5229 g/mol |
Número MDL:
MFCD01346689 Núm. CAS: 155-12-4 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...