2-Chloropyrimidine-5-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2-chloro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

C3436-1G C3436-5G
TCIAC3436-1GEA 56 EUR
TCIAC3436-1G TCIAC3436-5G
2-Chloropyrimidine-5-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2-Chloropyrimidine-5-boronic acid pinacol ester
Formula: C₁₀H₁₄BClN₂O₂
MW: 240,494 g/mol
Número MDL: MFCD11856048
Núm. CAS: 1003845-08-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9faddc22fb8d65a2efaf78a1d0b278c8fc67c11c&vid=9faddc22fb8d65a2efaf78a1d0b278c8fc67c11c&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 65.0 to 69.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR