(6-Chloro-1H-benzotriazol-1-yloxy)tripyrrolidinophosphonium Hexafluorophosphate ≥98.0% (by HPLC)
Proveedor: TCI
|
Atención
|
Sinónimos:
((6-Chloro-1H-benzo[d][1,2,3]triazol-1-yl)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate, TPTDP
C3461-5G
C3461-25G
TCIAC3461-5GEA
38
EUR
TCIAC3461-5G
TCIAC3461-25G
(6-Chloro-1H-benzotriazol-1-yloxy)tripyrrolidinophosphonium Hexafluorophosphate ≥98.0% (by HPLC)
(6-Chloro-1H-benzotriazol-1-yloxy)tripyrrolidinophosphonium Hexafluorophosphate
Formula:
C₁₈H₂₇ClF₆N₆OP₂ MW: 554,84 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD14560589 Núm. CAS: 893413-42-8 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...