Calcium bis(fluorosulphonyl)imide ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

C3476-1G C3476-5G
TCIAC3476-1GEA 149 EUR
TCIAC3476-1G TCIAC3476-5G
Calcium bis(fluorosulphonyl)imide ≥98.0% (by titrimetric analysis)
Calcium bis(fluorosulphonyl)imide
Formula: CaF₄N₂O₈S₄
MW: 400,32 g/mol
Núm. CAS: 1906900-37-5
UN: 1759
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76c56b506e498087548c84250046a8be5b3e2a19&vid=76c56b506e498087548c84250046a8be5b3e2a19&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR