Carboxin ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Atención
|
C3482-25G
C3482-5G
TCIAC3482-25GEA
342
EUR
TCIAC3482-25G
TCIAC3482-5G
Carboxin ≥98.0% (by HPLC, total nitrogen)
Carboxin
Formula:
C₁₂H₁₃NO₂S |
Número MDL:
MFCD00055403 Núm. CAS: 5234-68-4 EINECS: 226-031-1 UN: 3077 ADR: 9,III Índice Merck: 14,01827 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 89.0 to 93.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...