Clevidipine ≥98.0% (by HPLC)
Proveedor: TCI
Sinónimos:
3-((Butyryloxy)methyl) 5-methyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, 4-(2,3-Dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylic Aicd 3-[(Butyryloxy)methyl] 5-Methyl ester, 3-[(Butyryloxy)methyl] 5-Methyl 4-(2,3-Dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
C3503-1G
C3503-250MG
TCIAC3503-1GEA
313
EUR
TCIAC3503-1G
TCIAC3503-250MG
Clevidipine ≥98.0% (by HPLC)
Clevidipine
Formula:
C₂₁H₂₃Cl₂NO₆ MW: 456,32 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00940070 Núm. CAS: 167221-71-8 Índice Merck: 14,02352 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 96.0 % |
Melting point | 141.0 to 145.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...