4-(3-Chlorophenyl)-2,6-diphenylpyrimidine ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Atención

C3519-1G C3519-5G
TCIAC3519-1GEA 53 EUR
TCIAC3519-1G TCIAC3519-5G
4-(3-Chlorophenyl)-2,6-diphenylpyrimidine ≥98.0% (by HPLC, total nitrogen)
4-(3-Chlorophenyl)-2,6-diphenylpyrimidine
Formula: C₂₂H₁₅ClN₂
MW: 342,83 g/mol
Número MDL: MFCD29477687
Núm. CAS: 1536209-87-6

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 144.0 to 148.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR