Cyclohexanemethylamine hydrobromide ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
(Aminomethyl)cyclohexane hydrobromide, Cyclohexylmethylamine hydrobromide, Cyclohexanemethylammonium bromide
C3531-1G
C3531-5G
TCIAC3531-1GEA
37
EUR
TCIAC3531-1G
TCIAC3531-5G
Cyclohexanemethylamine hydrobromide ≥98.0% (by titrimetric analysis)
Cyclohexanemethylamine hydrobromide
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...