Clodinafop-propargyl ≥98.0% (by total nitrogen basis)
Proveedor: TCI
|
Atención
|
Sinónimos:
(R)-2-[4-(5-Chloro-3-fluoro-2-pyridinyloxy)phenoxy]propanoic acid 2-Propynyl ester, 2-Propynyl (R)-2-[4-(5-Chloro-3-fluoro-2-pyridinyloxy)phenoxy]propanoate
C3552-5G
C3552-25G
TCIAC3552-5GEA
77
EUR
TCIAC3552-5G
TCIAC3552-25G
Clodinafop-propargyl ≥98.0% (by total nitrogen basis)
Clodinafop-propargyl
Formula:
C₁₇H₁₃ClFNO₄ MW: 349,74 g/mol |
Número MDL:
MFCD01632328 Núm. CAS: 105512-06-9 UN: 3077 ADR: 9,III Índice Merck: 14,02370 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 57.0 to 61.0 deg-C |
Specific rotation [a]20/D | +45.0 to +50.0 deg (C=2, Acetone) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...