3-Chloro-N,N-diethylpropan-1-amine ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
C3578-25G
C3578-5G
TCIAC3578-25GEA
69
EUR
TCIAC3578-25G
TCIAC3578-5G
3-Chloro-N,N-diethylpropan-1-amine ≥98.0% (by GC, titration analysis)
3-Chloro-N,N-diethylpropan-1-amine
Formula:
C₇H₁₆ClN Pto. Ebullición: 160°C |
Número MDL:
MFCD00039385 Núm. CAS: 104-77-8 EINECS: 203-235-9 UN: 2734 ADR: 8,II |
Resultados test especificaciones
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...