2-Chloro-3-methyl-5-pyridineboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: 2-Chloro-3-methylpyridine-5-boronic acid, 6-Chloro-5-methylpyridine-3-boronic acid, (6-Chloro-5-methylpyridin-3-yl)boronic acid

C3588-1G C3588-5G
TCIAC3588-1GEA 37 EUR
TCIAC3588-1G TCIAC3588-5G
2-Chloro-3-methyl-5-pyridineboronic acid (contains varying amounts of Anhydride)
2-Chloro-3-methyl-5-pyridineboronic acid
Formula: C₆H₇BClNO₂
Número MDL: MFCD03411564
Núm. CAS: 1003043-40-0

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 111.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR