4'-Chlorobiphenyl-4-ylboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: 4'-Chloro-4-biphenylboronic acid, (4'-Chloro-[1,1'-biphenyl]-4-yl)boronic acid

C3614-1G C3614-5G
TCIAC3614-1GEA 44 EUR
TCIAC3614-1G TCIAC3614-5G
4'-Chlorobiphenyl-4-ylboronic acid (contains varying amounts of Anhydride)
4'-Chlorobiphenyl-4-ylboronic acid
Formula: C₁₂H₁₀BClO₂
MW: 232,4700 g/mol
Número MDL: MFCD08544389
Núm. CAS: 364044-44-0

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) 98.0 to 107.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR