Cyanomethyl Dodecyl trithiocarbonate ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Atención
|
Sinónimos:
Cyanomethyl dodecyl carbonotrithioate
C3626-1G
C3626-5G
TCIAC3626-1GEA
234
EUR
TCIAC3626-1G
TCIAC3626-5G
Cyanomethyl Dodecyl trithiocarbonate ≥98.0% (by HPLC, total nitrogen)
Cyanomethyl Dodecyl trithiocarbonate
Formula:
C₁₅H₂₇NS₃ MW: 317,57 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD11045090 Núm. CAS: 796045-97-1 |
Resultados test especificaciones
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 29.0 to 33.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...