Decenylsuccinic anhydride ≥95.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
D0030-25G
D0030-100G
TCIAD0030-25GEA
49
EUR
TCIAD0030-25G
TCIAD0030-100G
Decenylsuccinic anhydride ≥95.0% (by titrimetric analysis)
Decenylsuccinic anhydride
Formula:
C₁₄H₂₂O₃ Pto. de inflamación: 174°C |
Número MDL:
MFCD00014547 Núm. CAS: 25447-83-0 EINECS: 246-994-1 |
Resultados test especificaciones
Appearance | Light orange to Yellow to Green clear liquid |
Purity (Morpholine Method) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...