Dehydroepiandrosterone Acetate ≥99.0% (por GC)
Proveedor: TCI
Sinónimos:
trans-Dehydroandrosterone Acetate, Dehydroisoandrosterone Acetate
D0045-25G
D0045-5G
TCIAD0045-25GEA
130
EUR
TCIAD0045-25G
TCIAD0045-5G
Dehydroepiandrosterone Acetate ≥99.0% (por GC)
Dehydroepiandrosterone Acetate
Formula:
C₂₁H₃₀O₃ MW: 330.47 g/mol |
Número MDL:
MFCD00021119 Núm. CAS: 853-23-6 EINECS: 212-714-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 169.0 to 172.0 deg-C |
Specific rotation [a]20/D | +3.0 to +6.0 deg (C=2, EtOH) |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...