Sodium dibenzyldithiocarbamate hydrate ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
Dibenzyldithiocarbamic Acid Sodium Salt
D0156-25G
D0156-5G
TCIAD0156-25GEA
343
EUR
TCIAD0156-25G
TCIAD0156-5G
Sodium dibenzyldithiocarbamate hydrate ≥98.0% (by titrimetric analysis)
Sodium dibenzyldithiocarbamate hydrate
Formula:
C₁₅H₁₅NS₂ MW: 295.39 g/mol |
Número MDL:
MFCD00066462 Núm. CAS: 55310-46-8 EINECS: 259-587-9 |
|
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (Iodometric Titration) | min. 98.0 % (calcd.on anh.substance) |
Solubility in EtOH | almost transparency |
Water (Vaporization methoed) | max. 27.0 % |
Sensitiveness | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...