3,5-Dibromo-L-tyrosine ≥96.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: H-Tyr(3,5-DiBr)-OH, H-3,5-Dibromo-Tyr-OH

D0213-1G D0213-5G
TCIAD0213-1GEA 26 EUR
TCIAD0213-1G TCIAD0213-5G
3,5-Dibromo-L-tyrosine ≥96.0% (by titrimetric analysis)
3,5-Dibromo-L-tyrosine
Formula: C₉H₉Br₂NO₃
Número MDL: MFCD00037204
Núm. CAS: 300-38-9
EINECS: 206-091-5
Índice Merck: 14,03030
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a72b9915faf77fcf3b8ddad064496ada0a341d98&vid=c0757b547072e82e759e9fca7c6f0001c6a9bae3&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 96.0 %
Specific rotation [a]20/D +0.5 to +2.0 deg (C=5, 1mol/L HCl)
Solubility in 1mol/L HCl almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR