Dichloramine B ≥95.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

D0317-25G D0317-5G
TCIAD0317-25GEA 203 EUR
TCIAD0317-25G TCIAD0317-5G
Dichloramine B ≥95.0% (by titrimetric analysis)
Dichloramine B
Formula: C₆H₅Cl₂NO₂S
Número MDL: MFCD00025025
Núm. CAS: 473-29-0
EINECS: 207-461-9
UN: 1479
ADR: 5.1,I

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Green powder to crystal
Purity (Iodometric Titration) min. 95.0 %
Melting point 70.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR