Guanylurea phosphate ≥98.0% (by titrimetric analysis)

Proveedor: TCI

D0432-25G D0432-500G
TCIAD0432-25GEA 16 EUR
TCIAD0432-25G TCIAD0432-500G
Guanylurea phosphate ≥98.0% (by titrimetric analysis)
Guanylurea phosphate
Formula: C₂H₆N₄O·H₃PO₄
Número MDL: MFCD00065097
Núm. CAS: 36897-89-9
EINECS: 253-264-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR