N-(2-Diethylaminoethyl)-1-naphthylamine oxalate ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: N,N-Diethyl-N'-1-naphthylethylenediamine Oxalate, N1,N1-Diethyl-N2-(naphthalen-1-yl)ethane-1,2-diamine oxalate, Tsuda's Reagent

D0469-1G D0469-25G D0469-5G
TCIAD0469-1GEA 16 EUR
TCIAD0469-1G TCIAD0469-25G TCIAD0469-5G
N-(2-Diethylaminoethyl)-1-naphthylamine oxalate ≥98.0% (by titrimetric analysis)
N-(2-Diethylaminoethyl)-1-naphthylamine oxalate
Formula: C₁₆H₂₄N₂O₄
MW: 332,40 g/mol
Número MDL: MFCD00060156
Núm. CAS: 29473-53-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Water almost transparency
Sensitiveness to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR