Hexestrol ≥98.0% (por GC)
Proveedor: TCI
|
Peligro
|
Sinónimos:
4,4'-(1,2-Diethylethylene)diphenol, rel-4,4'-((3R,4S)-Hexane-3,4-diyl)diphenol
D0494-1G
D0494-10G
TCIAD0494-1GEA
46
EUR
TCIAD0494-1G
TCIAD0494-10G
Hexestrol ≥98.0% (por GC)
Hexestrol
Formula:
C₁₈H₂₂O₂ MW: 270,37 g/mol |
Número MDL:
MFCD00068996 Núm. CAS: 84-16-2 EINECS: 201-518-1 Índice Merck: 14,04701 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 184.0 to 188.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...