N,N-Diethylglycinesodium salt ≥97.0% (by titrimetric analysis)

Proveedor: TCI

D0509-100G D0509-25G
TCIAD0509-100GEA 313 EUR
TCIAD0509-100G TCIAD0509-25G
N,N-Diethylglycinesodium salt ≥97.0% (by titrimetric analysis)
N,N-Diethylglycinesodium salt
Formula: C₆H₁₃NO₂
MW: 153.16 g/mol
Número MDL: MFCD00053497
Núm. CAS: 5426-55-1

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR