Ethyl red ≥97.0% (by titrimetric analysis) not cyanin dyes

Proveedor: TCI

Sinónimos: 4-(Diethylamino)azobenzene-2'-carboxylic Acid, Diethyl Red

D0524-1G D0524-25G
TCIAD0524-1GEA 16 EUR
TCIAD0524-1G TCIAD0524-25G
Ethyl red ≥97.0% (by titrimetric analysis) not cyanin dyes
Ethyl red
Formula: C₁₇H₁₉N₃O₂
MW: 297,36 g/mol
Número MDL: MFCD00002427
Núm. CAS: 76058-33-8

Order Now

Resultados test especificaciones

Appearance Orange to Brown to Dark purple powder to crystal
Purity (Neutralization titration) min. 97.0 %
Melting point 144.0 to 147.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR