Sodium-2,4-dimethylbenzenesulphonate monohydrate ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: m-Xylene-4-sulphonic acid Sodium Salt, Sodium m-Xylene-4-sulphonate, 2,4-Dimethylbenzenesulphonic acid Sodium Salt

D0682-25G D0682-500G
TCIAD0682-25GEA 16 EUR
TCIAD0682-25G TCIAD0682-500G
Sodium-2,4-dimethylbenzenesulphonate monohydrate ≥98.0% (by titrimetric analysis)
Sodium-2,4-dimethylbenzenesulphonate monohydrate
Formula: C₈H₁₀O₃S
MW: 208,21 g/mol
Número MDL: MFCD00041883
Núm. CAS: 142063-30-7
EINECS: 212-567-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0c594a3b45055137fa32b377a8af31be254a7cc&vid=438cc84fe9b7e483da78b855ac7ca33bfb432efc&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 % (as Monohydrate)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR