Dimethylammonium dimethyldithiocarbamate ≥95.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: Dimethyldithiocarbamic Acid Dimethylammonium Salt

D0715-25G D0715-5G
TCIAD0715-25GEA 146 EUR
TCIAD0715-25G TCIAD0715-5G
Dimethylammonium dimethyldithiocarbamate ≥95.0% (by titrimetric analysis)
Dimethylammonium dimethyldithiocarbamate
Formula: C₅H₁₄N₂S₂
MW: 166,30 g/mol
Número MDL: MFCD00067267
Núm. CAS: 598-64-1
EINECS: 209-945-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=891f61fe10bb434e2b2ff323422a77f286ec8a42&vid=891f61fe10bb434e2b2ff323422a77f286ec8a42&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (Iodometric Titration) min. 95.0 %
Melting point 129.0 to 133.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR