1,3-Bis(hydroxymethyl)urea ≥97.0% (by total nitrogen basis)

Proveedor: TCI
Atención

Sinónimos: N,N'-Bis(hydroxymethyl)urea, N,N'-Dimethylolurea

D0767-25G D0767-500G
TCIAD0767-25GEA 16 EUR
TCIAD0767-25G TCIAD0767-500G
1,3-Bis(hydroxymethyl)urea ≥97.0% (by total nitrogen basis)
1,3-Bis(hydroxymethyl)urea
Formula: C₃H₈N₂O₃
MW: 120,11 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00014414
Núm. CAS: 140-95-4
EINECS: 205-444-0
Índice Merck: 14,06968

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) 97.0 to 103.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR