2,2'-Dinaphthyl ether ≥99.0% (por GC)
Proveedor: TCI
D0811-10G
D0811-25G
TCIAD0811-10GEA
129
EUR
TCIAD0811-10G
TCIAD0811-25G
2,2'-Dinaphthyl ether ≥99.0% (por GC)
2,2'-Dinaphthyl ether
Formula:
C₂₀H₁₄O MW: 270.33 g/mol Pto. Ebullición: 250°C |
Número MDL:
MFCD00015447 Núm. CAS: 613-80-9 EINECS: 210-356-0 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 103.0 to 107.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...