Diphenylamine sulphate ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
D0875-25G
D0875-500G
TCIAD0875-25GEA
51
EUR
TCIAD0875-25G
TCIAD0875-500G
Diphenylamine sulphate ≥98.0% (by HPLC, titration analysis)
Diphenylamine sulphate
Formula:
C₁₂H₁₁N |
Número MDL:
MFCD00042016 Núm. CAS: 587-84-8 EINECS: 209-605-6 Índice Merck: 14,03317 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 147.0 to 152.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...