Methylenediphenyl-4,4'-diisocyanate ≥97.0% (by titrimetric analysis)
Proveedor: TCI
|
Peligro
|
D0897-25G
D0897-500G
TCIAD0897-25GEA
16
EUR
TCIAD0897-25G
TCIAD0897-500G
Methylenediphenyl-4,4'-diisocyanate ≥97.0% (by titrimetric analysis)
Methylenediphenyl-4,4'-diisocyanate
Formula:
C₁₅H₁₀N₂O₂ Pto. Ebullición: 184°C Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00036131 Núm. CAS: 101-68-8 EINECS: 202-966-0 UN: 2206 ADR: 6.1,II |
Resultados test especificaciones
Appearance | White to Almost white powder to lump |
Purity (Butylamine Method) | min. 97.0 % |
Melting point | 40.0 to 50.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...