1,4-Diphenylsemicarbazide ≥98.0% (by HPLC)
Proveedor: TCI
|
Atención
|
D0913-25G
D0913-5G
TCIAD0913-25GEA
203
EUR
TCIAD0913-25G
TCIAD0913-5G
1,4-Diphenylsemicarbazide ≥98.0% (by HPLC)
1,4-Diphenylsemicarbazide
Formula:
C₁₃H₁₃N₃O |
Número MDL:
MFCD00020635 Núm. CAS: 621-12-5 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 177.0 to 183.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...