Nε-(2,4-dinitrophenyl)-L-lysine hydrochloride ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: H-Lys(Dnp)-OH.HCl, Nε-Dnp-L-lysine Hydrochloride

D1023-1G D1023-100MG
TCIAD1023-1GEA 158 EUR
TCIAD1023-1G TCIAD1023-100MG
Nε-(2,4-dinitrophenyl)-L-lysine hydrochloride ≥98.0% (by HPLC)
Nepsilon-(2,4-dinitrophenyl)-L-lysine hydrochloride
Formula: C₁₂H₁₆N₄O₆
MW: 348,74 g/mol
Número MDL: MFCD00036383
Núm. CAS: 14401-10-6
EINECS: 238-366-0

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 94.0 %
Specific rotation [a]20/D +18.0 to +20.0 deg (C=2, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR