N-(2,4-Dinitrophenyl)-L-leucine ≥99.0% (by titrimetric analysis)
Proveedor: TCI
|
Peligro
|
D1033-1G
D1033-100MG
TCIAD1033-1GEA
115
EUR
TCIAD1033-1G
TCIAD1033-100MG
N-(2,4-Dinitrophenyl)-L-leucine ≥99.0% (by titrimetric analysis)
N-(2,4-Dinitrophenyl)-L-leucine
Formula:
C₁₂H₁₅N₃O₆ |
Número MDL:
MFCD00038102 Núm. CAS: 1655-57-8 UN: 1325 ADR: 4.1,II |
Resultados test especificaciones
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 96.0 to 100.0 deg-C |
Specific rotation [a]20/D | +54.0 to +59.0 deg (C=0.5, NaHCO3 aq. (4%)) |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...