N-(2,4-Dinitrophenyl)-L-valine ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

Sinónimos: N-Dnp-L-valine, Dnp-Val-OH

D1043-1G D1043-100MG
TCIAD1043-1GEA 67 EUR
TCIAD1043-1G TCIAD1043-100MG
N-(2,4-Dinitrophenyl)-L-valine ≥98.0% (by HPLC, titration analysis)
N-(2,4-Dinitrophenyl)-L-valine
Formula: C₁₁H₁₃N₃O₆
MW: 283.24 g/mol
Número MDL: MFCD00038096
Núm. CAS: 1694-97-9
EINECS: 216-909-2
UN: 1325
ADR: 4.1 II

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 129.0 to 133.0 deg-C
Specific rotation [a]20/D -24.0 to -28.0 deg (C=1, AcOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR