1,4-Bis(2-hydroxyethoxy)benzene ≥95.0% (por GC)
Proveedor: TCI
Sinónimos:
Hydroquinone Di(2-hydroxyethyl) Ether, 1,4-Di(2-hydroxyethoxy)benzene, Hydroquinone Bis(2-hydroxyethyl) Ether
D1065-25G
D1065-500G
TCIAD1065-25GEA
17
EUR
TCIAD1065-25G
TCIAD1065-500G
1,4-Bis(2-hydroxyethoxy)benzene ≥95.0% (por GC)
1,4-Bis(2-hydroxyethoxy)benzene
Formula:
C₁₀H₁₄O₄ MW: 198,22 g/mol |
Número MDL:
MFCD00002861 Núm. CAS: 104-38-1 EINECS: 203-197-3 |
Resultados test especificaciones
Appearance | White to Light yellow to Dark green powder to crystaline |
Purity (GC) | min. 95.0 % |
Melting point | 103.0 to 106.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...