Bis(2-benzamidophenyl)disulphide ≥98.0% (by total nitrogen basis)
Proveedor: TCI
|
Atención
|
D1102-25G
D1102-500G
TCIAD1102-25GEA
17
EUR
TCIAD1102-25G
TCIAD1102-500G
Bis(2-benzamidophenyl)disulphide ≥98.0% (by total nitrogen basis)
Bis(2-benzamidophenyl)disulphide
Formula:
C₂₆H₂₀N₂O₂S₂ |
Número MDL:
MFCD00043806 Núm. CAS: 135-57-9 EINECS: 205-201-9 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 140.0 to 145.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...