DL-2,4-Diaminobutyric acid dihydrochloride ≥98.0% (by total nitrogen and titration analysis)

Proveedor: TCI
Atención

Sinónimos: H-DL-Dab·2HCl, H-DL-Dab.2HCl

D1175-1G D1175-25G D1175-5G
TCIAD1175-1GEA 86 EUR
TCIAD1175-1G TCIAD1175-25G TCIAD1175-5G
DL-2,4-Diaminobutyric acid dihydrochloride ≥98.0% (by total nitrogen and titration analysis)
DL-2,4-Diaminobutyric acid dihydrochloride
Formula: C₄H₁₀N₂O₂·₂HCl
MW: 191,1 g/mol
Número MDL: MFCD00012910
Núm. CAS: 65427-54-5
EINECS: 265-774-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 201.0 to 205.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR