1,3-Diphenylguanidine sulphate

Proveedor: TCI

D1348-25G D1348-500G
TCIAD1348-25GEA 66 EUR
TCIAD1348-25G TCIAD1348-500G
1,3-Diphenylguanidine sulphate
1,3-Diphenylguanidine sulphate
Formula: C₁₃H₁₃N₃
MW: 520.61 g/mol
Número MDL: MFCD00060241
Núm. CAS: 32514-47-9

Order Now

Resultados test especificaciones

Appearance White powder to crystal
Purity (with Total Nitrogen) min. 95.0 % (calcd.on dried substance)
Solubility in hot Water almost transparency
Drying loss max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR