Clonidine hydrochloride ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Peligro
|
D1353-1G
D1353-5G
TCIAD1353-1GEA
36
EUR
TCIAD1353-1G
TCIAD1353-5G
Clonidine hydrochloride ≥98.0% (by HPLC, titration analysis)
Clonidine hydrochloride
Formula:
C₉H₉Cl₂N₃ Pto. de fusión: 312°C |
Número MDL:
MFCD00036705 Núm. CAS: 4205-91-8 EINECS: 224-121-5 UN: 2811 ADR: 6.1,I Índice Merck: 14,02390 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...