(S)-2,3-Diaminopropanoic acid hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
H-Dap-OH.HCl, H-Dap-OH·HCl, L-3-Aminoalanine Hydrochloride, (S)-2,3-Diaminopropionic acid monohydrochloride
D1377-1G
D1377-100MG
TCIAD1377-1GEA
100
EUR
TCIAD1377-1G
TCIAD1377-100MG
(S)-2,3-Diaminopropanoic acid hydrochloride ≥98.0% (by total nitrogen and titration analysis)
(S)-2,3-Diaminopropanoic acid hydrochloride
Formula:
C₃H₈N₂O₂·HCl MW: 140,6 g/mol |
Número MDL:
MFCD00065497 Núm. CAS: 1482-97-9 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Specific rotation [a]20/D | +22.0 to +26.0 deg (C=2, 1mol/L HCl) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...