Dicyclohexylamine nitrite ≥97.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

Sinónimos: N,N-Dicyclohexylammonium nitrite

D1392-25G D1392-500G
TCIAD1392-25GEA 16 EUR
TCIAD1392-25G TCIAD1392-500G
Dicyclohexylamine nitrite ≥97.0% (by titrimetric analysis)
Dicyclohexylamine nitrite
Formula: C₁₂H₂₃N
MW: 228,34 g/mol
Pto. de fusión: 183°C
Número MDL: MFCD00013128
Núm. CAS: 3129-91-7
EINECS: 221-515-9
UN: 2687
ADR: 4.1,III

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR