Methyl dihydrojasmonate (cis and trans mixture) cis- and trans-mixture ≥95.0% (por GC)
Proveedor: TCI
Sinónimos:
2-Amylcyclopentan-1-one-3-acetic Acid Methyl Ester, Methyl 3-Oxo-2-pentyl-1-cyclopentaneacetate, Methyl dihydrojasmonate, Methyl 2-(3-oxo-2-pentylcyclopentyl)acetate, Dihydrojasmonic Acid Methyl Ester
D1431-25ML
D1431-100ML
TCIAD1431-25MLEA
25
EUR
TCIAD1431-25ML
TCIAD1431-100ML
Methyl dihydrojasmonate (cis and trans mixture) cis- and trans-mixture ≥95.0% (por GC)
Methyl dihydrojasmonate (cis and trans mixture)
Formula:
C₁₃H₂₂O₃ Pto. Ebullición: 110°C Pto. de inflamación: 156°C |
Número MDL:
MFCD00151188 Núm. CAS: 24851-98-7 EINECS: 246-495-9 Índice Merck: 14,06052 |
Resultados test especificaciones
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % (total of isomer) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...