N,N-Diethylglycine methyl ester ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
D1443-25G
D1443-500G
D1443-5G
TCIAD1443-25GEA
61
EUR
TCIAD1443-25G
TCIAD1443-500G
TCIAD1443-5G
N,N-Diethylglycine methyl ester ≥98.0% (by GC, titration analysis)
N,N-Diethylglycine methyl ester
Formula:
C₇H₁₅NO₂ MW: 145,20 g/mol Pto. Ebullición: 86°C Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00059368 Núm. CAS: 30280-35-4 UN: 1993 ADR: 3,III |
Resultados test especificaciones
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...